(1,3-Dioxoisoindolin-2-yl) 2-(4-isobutylphenyl)propanoate 97%

Supplier: Ambeed
Warning

A635396-1G A635396-250MG A635396-5G
76650-454EA 34 USD
76650-454 76650-456 76650-458
(1,3-Dioxoisoindolin-2-yl) 2-(4-isobutylphenyl)propanoate 97%
(1,3-Dioxoisoindolin-2-yl) 2-(4-isobutylphenyl)propanoate

Precaución: for Research Use Only

Formula: C₂₁H₂₁NO₄
CAS Number: 2140889-61-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR