N-Fmoc-N-(2-phenylethyl)-glycine 98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-N-(2-PhenylEt)-Gly-OH

A634801-1G A634801-250MG A634801-5G
76650-446EA 49 USD
76650-446 76650-448 77783-543
N-Fmoc-N-(2-phenylethyl)-glycine 98%
N-Fmoc-N-(2-phenylethyl)-glycine

Precaución: for Research Use Only

Formula: C₂₅H₂₃NO₄
MW: 401.4544 g/mol
MDL Number: MFCD30187632
CAS Number: 540483-58-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR