9-(2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-9H-carbazole 95%

Supplier: Ambeed
Warning

Synonyms: 9-[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-9H-carbazole

A633993-100MG A633993-1G A633993-250MG A633993-5G
76650-426EA 22.28 USD
76650-426 76650-428 76650-430 76650-432
9-(2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-9H-carbazole 95%
9-(2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-9H-carbazole

Precaución: for Research Use Only

Formula: C₂₄H₂₄BNO₂
MW: 369.2639 g/mol
MDL Number: MFCD30737495
CAS Number: 1357634-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR