6-(4-isopropylphenyl)-2-(4-((4-methylpiperazin-1-yl)methyl)piperidin-1-yl)imidazo[2,1-b][1,3,4]thiadiazole 98%

Supplier: Ambeed
Warning

Synonyms: E260

A633521-10MG A633521-1MG A633521-25MG A633521-5MG
76650-412EA 91.24 USD
76650-412 76650-414 76650-416 76650-418
6-(4-isopropylphenyl)-2-(4-((4-methylpiperazin-1-yl)methyl)piperidin-1-yl)imidazo[2,1-b][1,3,4]thiadiazole 98%
6-(4-isopropylphenyl)-2-(4-((4-methylpiperazin-1-yl)methyl)piperidin-1-yl)imidazo[2,1-b][1,3,4]thiadiazole

Precaución: for Research Use Only

Formula: C₂₄H₃₄N₆S
MW: 438.63 g/mol
MDL Number: MFCD31657143
CAS Number: 1241537-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR