3,3-Difluoropropan-1-ol 98%

Supplier: Ambeed
Danger

A632898-1G A632898-250MG A632898-5G
76650-404EA 346.92 USD
76650-404 77749-291 77749-292
3,3-Difluoropropan-1-ol 98%
3,3-Difluoropropan-1-ol

Precaución: for Research Use Only

Formula: C₃H₆F₂O
MW: 96.08 g/mol
MDL Number: MFCD22413846
CAS Number: 461-52-9
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR