N-Fmoc-6-Fluoro-L-tryptophan 95%

Supplier: Ambeed
Warning

A632425-100MG A632425-1G A632425-5G A632425-250MG
76650-402EA 20 USD
76650-402 77551-908 77783-534 77783-533
N-Fmoc-6-Fluoro-L-tryptophan 95%
N-Fmoc-6-Fluoro-L-tryptophan

Precaución: for Research Use Only

Formula: C₂₆H₂₁FN₂O₄
MDL Number: MFCD29917530
CAS Number: 908847-01-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR