UDP-α-D-N-Acetylglucosamine Disodium Salt 98%

Supplier: Ambeed
Warning

A632171-50MG A632171-1G A632171-250MG A632171-100MG A632171-10MG A632171-5MG
76650-400EA 158 USD
76650-400 77594-946 77598-636 77617-666 77617-668 77617-670
UDP-α-D-N-Acetylglucosamine Disodium Salt 98%
UDP-α-D-N-Acetylglucosamine Disodium Salt

Precaución: for Research Use Only

Formula: C₁₇H₂₅N₃Na₂O₁₇P₂
MDL Number: MFCD00799985
CAS Number: 91183-98-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR