Ethyl 4-bromopentanoate 97%

Supplier: Ambeed
Warning

A630551-250MG A630551-5G A630551-100MG
76650-378EA 20 USD
76650-378 76650-380 77783-526
Ethyl 4-bromopentanoate 97%
Ethyl 4-bromopentanoate

Precaución: for Research Use Only

Formula: C₇H₁₃BrO₂
MDL Number: MFCD16693250
CAS Number: 27126-42-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR