Methyl 3-bromo-1-methyl-1H-pyrazole-5-carboxylate 97%

Supplier: Ambeed
Warning

A630419-100MG A630419-1G A630419-5G A630419-250MG
76650-372EA 87 USD
76650-372 76650-374 76650-376 77598-628
Methyl 3-bromo-1-methyl-1H-pyrazole-5-carboxylate 97%
Methyl 3-bromo-1-methyl-1H-pyrazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₆H₇BrN₂O₂
MW: 219.0360 g/mol
MDL Number: MFCD28650794
CAS Number: 1784137-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR