2-(5-Methoxy-2-(trifluoromethyl)phenyl)acetic acid 97%

Supplier: Ambeed
Warning

A630239-1G A630239-100MG
76650-370EA 132.62 USD
76650-370 77622-400
2-(5-Methoxy-2-(trifluoromethyl)phenyl)acetic acid 97%
2-(5-Methoxy-2-(trifluoromethyl)phenyl)acetic acid

Precaución: for Research Use Only

Formula: C₁₀H₉F₃O₃
MW: 234.17 g/mol
MDL Number: MFCD13194424
CAS Number: 1214361-90-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR