3-(2-Bromophenyl)-9-phenyl-9H-carbazole 98%

Supplier: Ambeed
Warning

A630107-1G A630107-250MG A630107-5G
76650-362EA 29 USD
76650-362 76650-364 76650-366
3-(2-Bromophenyl)-9-phenyl-9H-carbazole 98%
3-(2-Bromophenyl)-9-phenyl-9H-carbazole

Precaución: for Research Use Only

Formula: C₂₄H₁₆BrN
MDL Number: MFCD29071836
CAS Number: 1190100-35-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR