1,10-Phenanthroline-4,7(1H,10H)-dione 95%

Supplier: Ambeed
Warning

A629586-1G A629586-250MG A629586-5G A629586-25G
76650-346EA 46 USD
76650-346 76650-348 76650-350 77749-285
1,10-Phenanthroline-4,7(1H,10H)-dione 95%
1,10-Phenanthroline-4,7(1H,10H)-dione

Precaución: for Research Use Only

Formula: C₁₂H₈N₂O₂
MDL Number: MFCD01548697
CAS Number: 87330-27-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR