5-Hydroxy-3-methyl-2(5H)-furanone 98%

Supplier: Ambeed
Warning

Synonyms: 5-Hydroxy-3-methyl-2,5-dihydrofuran-2-one, 5-Hydroxy-3-methylfuran-2(5H)-one

A629190-100MG A629190-250MG A629190-1G
76650-342EA 21 USD
76650-342 76650-344 77622-398
5-Hydroxy-3-methyl-2(5H)-furanone 98%
5-Hydroxy-3-methyl-2(5H)-furanone

Precaución: for Research Use Only

Formula: C₅H₆O₃
MW: 114.10 g/mol
MDL Number: MFCD15145822
CAS Number: 931-23-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR