5-Cyano-2-methylbenzenesulfonyl chloride 97%

Supplier: Ambeed
Danger

A628797-100MG A628797-1G A628797-250MG A628797-5G
76650-328EA 53.05 USD
76650-328 76650-330 76650-332 76650-334
5-Cyano-2-methylbenzenesulfonyl chloride 97%
5-Cyano-2-methylbenzenesulfonyl chloride

Precaución: for Research Use Only

Formula: C₈H₆ClNO₂S
MW: 215.6567 g/mol
MDL Number: MFCD09741164
CAS Number: 372198-49-7
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR