(2-Bromoethoxy)cyclohexane 95%

Supplier: Ambeed
Danger

Synonyms: Cyclohexane-PEG1-Br

A628310-1G A628310-250MG
76650-322EA 541.06 USD
76650-322 76650-324
(2-Bromoethoxy)cyclohexane 95%
(2-Bromoethoxy)cyclohexane

Precaución: for Research Use Only

Formula: C₈H₁₅BrO
MW: 207.1081 g/mol
MDL Number: MFCD09815847
CAS Number: 131665-94-6
UN: 3082
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR