2-Chloro-4-iodo-3-methylbenzonitrile 92%

Supplier: Ambeed
Warning

A628307-100MG A628307-1G A628307-250MG A628307-50MG
76650-314EA 117.76 USD
76650-314 76650-316 76650-318 76650-320
2-Chloro-4-iodo-3-methylbenzonitrile 92%
2-Chloro-4-iodo-3-methylbenzonitrile

Precaución: for Research Use Only

Formula: C₈H₅ClIN
MW: 277.489 g/mol
MDL Number: MFCD13192054
CAS Number: 757247-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR