2-Hydroxy-N-phenylacetamide 95%

Supplier: Ambeed
Warning

A628198-250MG A628198-100MG A628198-1G
76650-312EA 73 USD
76650-312 77573-042 77598-616
2-Hydroxy-N-phenylacetamide 95%
2-Hydroxy-N-phenylacetamide

Precaución: for Research Use Only

Formula: C₈H₉NO₂
MDL Number: MFCD00068778
CAS Number: 4746-61-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR