Allyl (2-aminoethyl)carbamate hydrochloride 95%

Supplier: Ambeed
Danger

Synonyms: Allyl (2-aminoethyl)carbamate HCl

A628024-250MG A628024-5G A628024-1G
76650-308EA 34 USD
76650-308 77598-614 77783-521
Allyl (2-aminoethyl)carbamate hydrochloride 95%
Allyl (2-aminoethyl)carbamate hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₃ClN₂O₂
MW: 180.63 g/mol
MDL Number: MFCD05865258
CAS Number: 1049722-41-9
UN: 1759
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR