(3-(Trifluoromethyl)pyridin-4-yl)methanol 95%

Supplier: Ambeed
Warning

A625887-100MG A625887-250MG
76650-302EA 47 USD
76650-302 76650-304
(3-(Trifluoromethyl)pyridin-4-yl)methanol 95%
(3-(Trifluoromethyl)pyridin-4-yl)methanol

Precaución: for Research Use Only

Formula: C₇H₆F₃NO
MDL Number: MFCD11848106
CAS Number: 1227598-69-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR