3-Benzyl-2-((Z)-((E)-5-(6-chloro-3-methylbenzo[d]thiazol-2(3H)-ylidene)-3-ethyl-4-oxothiazolidin-2-ylidene)methyl)thiazol-3-ium chloride 97%

Supplier: Ambeed
Warning

Synonyms: JG-98

A625683-10MG A625683-1MG A625683-25MG A625683-50MG A625683-5MG
76650-292EA 90.18 USD
76650-292 76650-294 76650-296 76650-298 76650-300
3-Benzyl-2-((Z)-((E)-5-(6-chloro-3-methylbenzo[d]thiazol-2(3H)-ylidene)-3-ethyl-4-oxothiazolidin-2-ylidene)methyl)thiazol-3-ium chloride 97%
3-Benzyl-2-((Z)-((E)-5-(6-chloro-3-methylbenzo[d]thiazol-2(3H)-ylidene)-3-ethyl-4-oxothiazolidin-2-ylidene)methyl)thiazol-3-ium chloride

Precaución: for Research Use Only

Formula: C₂₄H₂₁Cl₂N₃OS₃
MW: 534.54 g/mol
MDL Number: MFCD32067914
CAS Number: 1456551-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR