4-(7-Methoxyquinolin-4-yl)-2-methylphenol 98%

Supplier: Ambeed
Warning

Synonyms: CU-CPT-9a

A624532-100MG A624532-10MG A624532-1MG A624532-25MG A624532-50MG A624532-5MG
76650-274EA 253 USD
76650-274 76650-276 76650-278 76650-280 76650-282 76650-284
4-(7-Methoxyquinolin-4-yl)-2-methylphenol 98%
4-(7-Methoxyquinolin-4-yl)-2-methylphenol

Precaución: for Research Use Only

Formula: C₁₇H₁₅NO₂
MW: 265.31 g/mol
MDL Number: MFCD31689252
CAS Number: 2165340-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR