2-Chloro-7-fluoro-4H-pyrido[1,2-a]pyrimidin-4-one 98%

Supplier: Ambeed
Danger

A623749-1G A623749-250MG A623749-5G
76650-270EA 314.03 USD
76650-270 76650-272 77617-628
2-Chloro-7-fluoro-4H-pyrido[1,2-a]pyrimidin-4-one 98%
2-Chloro-7-fluoro-4H-pyrido[1,2-a]pyrimidin-4-one

Precaución: for Research Use Only

Formula: C₈H₄ClFN₂O
MW: 198.58 g/mol
MDL Number: MFCD28659052
CAS Number: 1449598-86-0
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR