Octan-2-yl 2-cyanoacrylate 95+% stabilized
Supplier: Ambeed
|
Warning
|
Synonyms:
Dermabond
Stabilization:
Stabilised with hydroquinone (HQ)
A617888-250MG
A617888-5G
A617888-1G
A617888-25G
76650-248EA
18.04
USD
76650-248
76650-250
77598-612
77617-610
Octan-2-yl 2-cyanoacrylate 95+% stabilized
Octan-2-yl 2-cyanoacrylate
Precaución: for Research Use Only
Formula:
C₁₂H₁₉NO₂ MW: 209.2848 g/mol |
MDL Number:
MFCD18072365 CAS Number: 133978-15-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...