3-Bromo-2-(methylsulfonyl)pyridine 95%

Supplier: Ambeed
Warning

A612421-1G A612421-250MG A612421-5G
76650-206EA 133.68 USD
76650-206 76650-208 77622-390
3-Bromo-2-(methylsulfonyl)pyridine 95%
3-Bromo-2-(methylsulfonyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₆BrNO₂S
MW: 236.08 g/mol
MDL Number: MFCD14702656
CAS Number: 1209459-95-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR