Methyl 2-cyano-6-methylbenzoate 95%

Supplier: Ambeed
Warning

A611380-1G A611380-5G A611380-250MG
76650-200EA 105 USD
76650-200 77749-259 77749-258
Methyl 2-cyano-6-methylbenzoate 95%
Methyl 2-cyano-6-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂
MDL Number: MFCD18399109
CAS Number: 127510-98-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR