1-Iodo-4-methoxy-2-methylbenzene 95%

Supplier: Ambeed
Warning

A610671-100MG A610671-1G A610671-250MG A610671-5G
76650-188EA 16 USD
76650-188 76650-190 76650-192 76650-194
1-Iodo-4-methoxy-2-methylbenzene 95%
1-Iodo-4-methoxy-2-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₉IO
MDL Number: MFCD11101045
CAS Number: 63452-69-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR