[2',4'-Bis(1,1-dimethylethyl)-6'-methoxy[1,1'-biphenyl]-2-yl]dicyclohexylphosphine 98%

Supplier: Ambeed
Warning

A610632-100MG A610632-1G A610632-250MG
76650-182EA 28.65 USD
76650-182 76650-184 76650-186
[2',4'-Bis(1,1-dimethylethyl)-6'-methoxy[1,1'-biphenyl]-2-yl]dicyclohexylphosphine 98%
[2',4'-Bis(1,1-dimethylethyl)-6'-methoxy[1,1'-biphenyl]-2-yl]dicyclohexylphosphine

Precaución: for Research Use Only

Formula: C₃₃H₄₉OP
MW: 492.7153 g/mol
MDL Number: MFCD30184625
CAS Number: 1848244-75-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR