Di-tert-butyl (R)-2-(aminomethyl)piperazine-1,4-dicarboxylate 97%
Supplier: Ambeed
|
Warning
|
A606550-250MG
A606550-5G
76650-136EA
97.61
USD
76650-136
77749-245
Di-tert-butyl (R)-2-(aminomethyl)piperazine-1,4-dicarboxylate 97%
Di-tert-butyl (R)-2-(aminomethyl)piperazine-1,4-dicarboxylate
Precaución: for Research Use Only
Formula:
C₁₅H₂₉N₃O₄ MW: 315.4085 g/mol |
MDL Number:
MFCD28501732 CAS Number: 1932411-32-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...