(3-Bromophenyl)diphenylphosphine oxide 98%

Supplier: Ambeed
Warning

A605593-1G A605593-5G
76650-130EA 33.95 USD
76650-130 76650-132
(3-Bromophenyl)diphenylphosphine oxide 98%
(3-Bromophenyl)diphenylphosphine oxide

Precaución: for Research Use Only

Formula: C₁₈H₁₄BrOP
MW: 357.19 g/mol
MDL Number: MFCD30533951
CAS Number: 10212-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR