6-Bromo-4-methylindoline-2,3-dione 95%

Supplier: Ambeed
Warning

A605556-100MG A605556-250MG
76650-120EA 128.37 USD
76650-120 76650-122
6-Bromo-4-methylindoline-2,3-dione 95%
6-Bromo-4-methylindoline-2,3-dione

Precaución: for Research Use Only

Formula: C₉H₆BrNO₂
MW: 240.0534 g/mol
MDL Number: MFCD31698218
CAS Number: 2384410-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR