(Dichloro((((((2R,3S,4R,5R)-3,4-dihydroxy-5-(6-((2-(methylthio)ethyl)amino)-2-((3,3,3-trifluoropropyl)thio)-9H-purin-9-yl)tetrahydrofuran-2-yl)methoxy)(hydroxy)phosphoryl)oxy)(hydroxy)phosphoryl)methyl)phosphonic acid tetrasodium salt 98%

Supplier: Ambeed
Warning

Synonyms: Cangrelor tetrasodium

A605340-10MG A605340-1MG A605340-25MG A605340-5MG
76650-110EA 65.78 USD
76650-110 76650-112 76650-114 76650-116
(Dichloro((((((2R,3S,4R,5R)-3,4-dihydroxy-5-(6-((2-(methylthio)ethyl)amino)-2-((3,3,3-trifluoropropyl)thio)-9H-purin-9-yl)tetrahydrofuran-2-yl)methoxy)(hydroxy)phosphoryl)oxy)(hydroxy)phosphoryl)methyl)phosphonic acid tetrasodium salt 98%
(Dichloro((((((2R,3S,4R,5R)-3,4-dihydroxy-5-(6-((2-(methylthio)ethyl)amino)-2-((3,3,3-trifluoropropyl)thio)-9H-purin-9-yl)tetrahydrofuran-2-yl)methoxy)(hydroxy)phosphoryl)oxy)(hydroxy)phosphoryl)methyl)phosphonic acid tetrasodium salt

Precaución: for Research Use Only

Formula: C₁₇H₂₁Cl₂F₃N₅Na₄O₁₂P₃S₂
MW: 864.2865 g/mol
MDL Number: MFCD14635359
CAS Number: 163706-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR