(3-Fluorobicyclo[1.1.1]pentan-1-yl)methanol 97%

Supplier: Ambeed
Warning

Synonyms: {3-fluorobicyclo[1.1.1]pentan-1-yl}methanol

A604361-50MG A604361-100MG
76650-102EA 307.67 USD
76650-102 77598-596
(3-Fluorobicyclo[1.1.1]pentan-1-yl)methanol 97%
(3-Fluorobicyclo[1.1.1]pentan-1-yl)methanol

Precaución: for Research Use Only

Formula: C₆H₉FO
MW: 116.133 g/mol
MDL Number: MFCD29037345
CAS Number: 262852-01-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR