5-Chloro-1H-pyrrolo[3,2-b]pyridine-3-carbonitrile ≥95%

Supplier: Ambeed
Warning

A603816-100MG A603816-250MG A603816-25MG
76650-096EA 254.62 USD
76650-096 76650-098 76650-100
5-Chloro-1H-pyrrolo[3,2-b]pyridine-3-carbonitrile ≥95%
5-Chloro-1H-pyrrolo[3,2-b]pyridine-3-carbonitrile

Precaución: for Research Use Only

Formula: C₈H₄ClN₃
MW: 177.5905 g/mol
MDL Number: MFCD18380894
CAS Number: 1260382-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR