5-Bromo-N-methylpyrimidine-2-carboxamide 95%

Supplier: Ambeed
Warning

A603524-100MG A603524-250MG
76650-092EA 23.34 USD
76650-092 76650-094
5-Bromo-N-methylpyrimidine-2-carboxamide 95%
5-Bromo-N-methylpyrimidine-2-carboxamide

Precaución: for Research Use Only

Formula: C₆H₆BrN₃O
MW: 216.0400 g/mol
MDL Number: MFCD20039121
CAS Number: 1346819-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR