Methyl 6-chloro-2-(difluoromethoxy)nicotinate 95%

Supplier: Ambeed
Warning

A603319-10G A603319-1G A603319-250MG
76650-086EA 1867.19 USD
76650-086 76650-088 76650-090
Methyl 6-chloro-2-(difluoromethoxy)nicotinate 95%
Methyl 6-chloro-2-(difluoromethoxy)nicotinate

Precaución: for Research Use Only

Formula: C₈H₆ClF₂NO₃
MW: 237.5880 g/mol
MDL Number: MFCD28751090
CAS Number: 1805519-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR