(1R,2S,5S)-rel-3-(tert-Butoxycarbonyl)-6,6-dimethyl-3-azabicyclo[3.1.0]hexane-2-carboxylic acid 98%
Supplier: Ambeed
|
Warning
|
A601366-100MG
A601366-1G
A601366-250MG
A601366-50MG
76650-064EA
114
USD
76650-064
76650-066
76650-068
76650-070
(1R,2S,5S)-rel-3-(tert-Butoxycarbonyl)-6,6-dimethyl-3-azabicyclo[3.1.0]hexane-2-carboxylic acid 98%
(1R,2S,5S)-rel-3-(tert-Butoxycarbonyl)-6,6-dimethyl-3-azabicyclo[3.1.0]hexane-2-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₃H₂₁NO₄ |
MDL Number:
MFCD31735161 CAS Number: 848777-73-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...