tert-Butyl 4,7-diazaspiro[2.5]octane-4-carboxylate hemioxalate 97%

Supplier: Ambeed
Warning

A600738-100MG A600738-1G A600738-250MG
76650-058EA 23.34 USD
76650-058 76650-060 76650-062
tert-Butyl 4,7-diazaspiro[2.5]octane-4-carboxylate hemioxalate 97%
tert-Butyl 4,7-diazaspiro[2.5]octane-4-carboxylate hemioxalate

Precaución: for Research Use Only

Formula: C₂₄H₄₂N₄O₈
MW: 514.6123 g/mol
MDL Number: MFCD31620825
CAS Number: 2173992-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR