4,6-Dichloro-1,3-dihydrofuro[3,4-c]pyridine 97%

Supplier: Ambeed
Warning

A600501-250MG A600501-1G A600501-100MG A600501-5G
76650-056EA 147 USD
76650-056 77598-594 77783-491 77783-492
4,6-Dichloro-1,3-dihydrofuro[3,4-c]pyridine 97%
4,6-Dichloro-1,3-dihydrofuro[3,4-c]pyridine

Precaución: for Research Use Only

Formula: C₇H₅Cl₂NO
MDL Number: MFCD31728404
CAS Number: 754992-21-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR