2-Iodo-N,N-dimethylaniline 95%
Supplier: Ambeed
|
Warning
|
A598030-1G
A598030-250MG
A598030-100MG
A598030-5G
76650-042EA
147
USD
76650-042
76650-044
77749-235
77749-236
2-Iodo-N,N-dimethylaniline 95%
2-Iodo-N,N-dimethylaniline
Precaución: for Research Use Only
Formula:
C₈H₁₀IN |
MDL Number:
MFCD20229908 CAS Number: 698-02-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...