1,1-Dimethyl-2-oxa-8-azaspiro[4.5]decane hydrochloride 97%

Supplier: Ambeed
Warning

A596377-100MG A596377-1G A596377-250MG
76650-026EA 168.69 USD
76650-026 76650-028 76650-030
1,1-Dimethyl-2-oxa-8-azaspiro[4.5]decane hydrochloride 97%
1,1-Dimethyl-2-oxa-8-azaspiro[4.5]decane hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₂₀ClNO
MW: 205.7249 g/mol
MDL Number: MFCD11035932
CAS Number: 1427425-91-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR