(S)-2-(6-Bromobenzo[d][1,3]dioxol-5-yl)-N-(4-cyanophenyl)-1-(1-cyclohexylethyl)-1H-benzo[d]imidazole-5-carboxamide 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-(6-Bromo-1,3-benzodioxol-5-yl)-N-(4-cyanophenyl)-1-[(1S)-1-cyclohexylethyl]-1H-benzimidazole-5-carboxamide, AZ3451, AZ 3451
A595701-1MG
A595701-5MG
76650-022EA
30
USD
76650-022
76650-024
(S)-2-(6-Bromobenzo[d][1,3]dioxol-5-yl)-N-(4-cyanophenyl)-1-(1-cyclohexylethyl)-1H-benzo[d]imidazole-5-carboxamide 97%
(S)-2-(6-Bromobenzo[d][1,3]dioxol-5-yl)-N-(4-cyanophenyl)-1-(1-cyclohexylethyl)-1H-benzo[d]imidazole-5-carboxamide
Precaución: for Research Use Only
Formula:
C₃₀H₂₇BrN₄O₃ MW: 571.4644 g/mol |
MDL Number:
MFCD31728469 CAS Number: 2100284-59-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...