Ethyl 3-amino-6-chloropyridazine-4-carboxylate 97%

Supplier: Ambeed
Warning

A594460-100MG A594460-1G A594460-5G
76650-018EA 36 USD
76650-018 76650-020 77598-592
Ethyl 3-amino-6-chloropyridazine-4-carboxylate 97%
Ethyl 3-amino-6-chloropyridazine-4-carboxylate

Precaución: for Research Use Only

Formula: C₇H₈ClN₃O₂
MW: 201.6100 g/mol
MDL Number: MFCD24642467
CAS Number: 1161847-32-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR