tert-Butyl 7-bromoheptanoate 97%

Supplier: Ambeed
Danger

A594121-100MG A594121-10G A594121-1G A594121-250MG A594121-5G A594121-25G
76650-008EA 19 USD
76650-008 76650-010 76650-012 76650-014 77598-590 77617-498
tert-Butyl 7-bromoheptanoate 97%
tert-Butyl 7-bromoheptanoate

Precaución: for Research Use Only

Formula: C₁₁H₂₁BrO₂
MDL Number: MFCD27924668
CAS Number: 51100-47-1
UN: 3272
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR