5-Methyl-2,3-dihydro-1H-indene-1-carboxylic acid 97%

Supplier: Ambeed
Warning

A593727-100MG A593727-250MG
76650-004EA 35 USD
76650-004 76650-006
5-Methyl-2,3-dihydro-1H-indene-1-carboxylic acid 97%
5-Methyl-2,3-dihydro-1H-indene-1-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₂
MDL Number: MFCD11219725
CAS Number: 956101-02-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR