2-Amino-6-azaspiro[3.4]octan-7-one hydrochloride 98%
Supplier: Ambeed
|
Warning
|
A592911-100MG
A592911-1G
A592911-250MG
A592911-50MG
76649-996EA
250
USD
76649-996
76649-998
76650-000
76650-002
2-Amino-6-azaspiro[3.4]octan-7-one hydrochloride 98%
2-Amino-6-azaspiro[3.4]octan-7-one hydrochloride
Precaución: for Research Use Only
Formula:
C₇H₁₃ClN₂O |
CAS Number:
2172063-36-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...