AZD-4635 97%

Supplier: Ambeed
Warning

A592542-10MG A592542-1MG A592542-25MG A592542-5MG A592542-100MG A592542-250MG A592542-50MG A592542-1G
76649-986EA 51 USD
76649-986 76649-988 76649-990 76649-992 77617-488 77617-490 77617-492 77749-219
AZD-4635 97%
AZD-4635

Precaución: for Research Use Only

Formula: C₁₅H₁₁ClFN₅
MDL Number: MFCD31618022
CAS Number: 1321514-06-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR