Lazertinib 97%

Supplier: Ambeed
Warning

Synonyms: N-(5-((4-(4-((Dimethylamino)methyl)-3-phenyl-1H-pyrazol-1-yl)pyrimidin-2-yl)amino)-4-methoxy-2-morpholinophenyl)acrylamide, N-[5-[[4-[4-[(dimethylamino)methyl]-3-phenylpyrazol-1-yl]pyrimidin-2-yl]amino]-4-methoxy-2-morpholin-4-ylphenyl]prop-2-enamide

A589050-10MG A589050-1MG A589050-50MG A589050-5MG
76649-978EA 59.42 USD
76649-978 76649-980 76649-982 76649-984
Lazertinib 97%
Lazertinib

Precaución: for Research Use Only

Formula: C₃₀H₃₄N₈O₃
MW: 554.66 g/mol
MDL Number: MFCD31728331
CAS Number: 1903008-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR