Adamantan-2-ylmethanamine hydrochloride 98%

Supplier: Ambeed
Warning

A586592-100MG A586592-1G A586592-250MG
76649-970EA 25 USD
76649-970 77783-478 77783-479
Adamantan-2-ylmethanamine hydrochloride 98%
Adamantan-2-ylmethanamine hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₂₀ClN
MDL Number: MFCD02208194
CAS Number: 79671-78-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR