5-Iodo-2,3-dihydro-1H-pyrrolo[2,3-b]pyridine 95%
Supplier: Ambeed
|
Warning
|
A584680-1G
A584680-5G
A584680-250MG
76649-964EA
124
USD
76649-964
77749-204
77749-203
5-Iodo-2,3-dihydro-1H-pyrrolo[2,3-b]pyridine 95%
5-Iodo-2,3-dihydro-1H-pyrrolo[2,3-b]pyridine
Precaución: for Research Use Only
Formula:
C₇H₇IN₂ |
MDL Number:
MFCD30528637 CAS Number: 2091464-45-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...