7-Fluoro-2-hydroxy-4H-pyrido[1,2-a]pyrimidin-4-one 97%
Supplier: Ambeed
|
Warning
|
A583042-1G
A583042-250MG
A583042-25G
A583042-5G
76649-954EA
17
USD
76649-954
76649-956
76649-958
76649-960
7-Fluoro-2-hydroxy-4H-pyrido[1,2-a]pyrimidin-4-one 97%
7-Fluoro-2-hydroxy-4H-pyrido[1,2-a]pyrimidin-4-one
Precaución: for Research Use Only
Formula:
C₈H₅FN₂O₂ MW: 180.1359 g/mol |
MDL Number:
MFCD28365090 CAS Number: 1449598-85-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...